Skip to Content
Merck
Fluorobenzene analytical standard

Fluorobenzene

Fluorobenzene analytical standard
Empirical Formula (Hill Notation):
C6H5F
CAS Number:
Molecular Weight:
96.10
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing