
(−)-Catechin 3-gallate

Synonym(s):
(−)-Catechin gallate, (2S,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-(3,4,5-trihydroxybenzoate)
Empirical Formula (Hill Notation):
C22H18O10
CAS Number:
Molecular Weight:
442.37
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing