Skip to Content
Merck
ANTI-MAGEB1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

ANTI-MAGEB1 antibody produced in rabbit

ANTI-MAGEB1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-CT3.1, Anti-Cancer/testis antigen 3.1, Anti-DAM10, Anti-DSS-AHC critical interval MAGE superfamily 10, Anti-MAGE-B1 antigen, Anti-MAGE-XP antigen, Anti-MAGEB1, Anti-MAGEB4, Anti-Melanoma-associated antigen B1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique