
Ammonium thiocyanate solution

Synonym(s):
Ammonium rhodanide solution
Linear Formula:
NH4SCN
CAS Number:
Molecular Weight:
76.12
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing