
Aluminium ammonium sulfate dodecahydrate

Synonym(s):
Aluminium ammonium sulfate dodecahydrate
Linear Formula:
NH4Al(SO4)2 · 12 H2O
CAS Number:
Molecular Weight:
453.33
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing