Skip to Content
Merck
All Photos(2)

Documents

472077

Sigma-Aldrich

2-(4-Hydroxyphenyl)-5-pyrimidinol

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O2
CAS Number:
Molecular Weight:
188.18
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

90%

form

powder or crystals

impurities

<10% water

mp

235-240 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc(cc1)-c2ncc(O)cn2

InChI

1S/C10H8N2O2/c13-8-3-1-7(2-4-8)10-11-5-9(14)6-12-10/h1-6,13-14H

InChI key

BKOPBDRWHKDPPN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service