Merck
  • Drug-Induced Self-Assembly of Modified Albumins as Nano-theranostics for Tumor-Targeted Combination Therapy.
Drug-Induced Self-Assembly of Modified Albumins as Nano-theranostics for Tumor-Targeted Combination Therapy.
ACS nano (2015-05-08)
Qian Chen, Xin Wang, Chao Wang, Liangzhu Feng, Yonggang Li, Zhuang Liu
ABSTRACT
Paclitaxel (PTX) can bind to human serum albumin (HSA) via hydrophobic interaction, forming Abraxane, which is a U.S. Food and Drug Administration (FDA) approved effective antitumor nanomedicine drug. Herein, the effective antitumor drug PTX is used to induce the self-assembly of HSA modified with either a photosensitizer chlorin e6 (Ce6), which at the same time serves as a chelating agent for Mn(2+) to enable magnetic resonance imaging, or acyclic Arg-Gly-Asp (cRGDyK) peptide that targets αvβ3-integrin overexpressed on tumor angiogenic endothelium. Two types of tumor-targeting theranostic nanoparticles are constructed, either by coassembly of both HSA-Ce6 and HSA-RGD simultaneously or by forming an HSA-Ce6@HSA-RGD core-shell structure, with the assistance of PTX-induced albumin aggregation. Such albumin-based nanoparticles on one hand could targetαvβ3-integrin, as evidenced by both in vitro and in vivo experiments, and on the other hand enable combined photodynamic/chemotherapy, which offers remarkably improved therapeutic efficacy to kill cancer in comparison to the respective monotherapies. Our work presents a new type of tumor-targeted multifunctional albumin-based nanoparticles by drug-induced self-assembly, which is a rather simple method without any sophisticated chemistry or materials engineering and is promising for multimodel imaging-guided combination therapy of cancer.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology