Merck
  • Biomarkers of one-carbon metabolism are associated with biomarkers of inflammation in women.
Biomarkers of one-carbon metabolism are associated with biomarkers of inflammation in women.
The Journal of nutrition (2014-03-22)
Clare Abbenhardt, Joshua W Miller, Xiaoling Song, Elissa C Brown, Ting-Yuan David Cheng, Mark H Wener, Yingye Zheng, Adetunji T Toriola, Marian L Neuhouser, Shirley A A Beresford, Karen W Makar, Lynn B Bailey, David R Maneval, Ralph Green, Joann E Manson, Linda Van Horn, Cornelia M Ulrich
ABSTRACT
Folate-mediated one-carbon metabolism is essential for DNA synthesis, repair, and methylation. Perturbations in one-carbon metabolism have been implicated in increased risk of some cancers and may also affect inflammatory processes. We investigated these interrelated pathways to understand their relation. The objective was to explore associations between inflammation and biomarkers of nutritional status and one-carbon metabolism. In a cross-sectional study in 1976 women selected from the Women's Health Initiative Observational Study, plasma vitamin B-6 [pyridoxal-5'-phosphate (PLP)], plasma vitamin B-12, plasma folate, and RBC folate were measured as nutritional biomarkers; serum C-reactive protein (CRP) and serum amyloid A (SAA) were measured as biomarkers of inflammation; and homocysteine and cysteine were measured as integrated biomarkers of one-carbon metabolism. Student's t, chi-square, and Spearman rank correlations, along with multiple linear regressions, were used to explore relations between biomarkers; additionally, we tested stratification by folic acid fortification period and multivitamin use. With the use of univariate analysis, plasma PLP was the only nutritional biomarker that was modestly significantly correlated with serum CRP and SAA (ρ = -0.22 and -0.12, respectively; P < 0.0001). Homocysteine (μmol/L) showed significant inverse correlations with all nutritional biomarkers (ranging from ρ = -0.30 to ρ = -0.46; all P < 0.0001). With the use of multiple linear regression, plasma PLP, RBC folate, homocysteine, and cysteine were identified as independent predictors of CRP; and PLP, vitamin B-12, RBC folate, and homocysteine were identified as predictors of SAA. When stratified by folic acid fortification period, nutrition-homocysteine correlations were generally weaker in the postfortification period, whereas associations between plasma PLP and serum CRP increased. Biomarkers of inflammation are associated with PLP, RBC folate, and homocysteine in women. The connection between the pathways needs to be further investigated and causality established. The trial is registered at clinicaltrials.gov as NCT00000611.
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
C Reactive Protein from human fluids, buffered aqueous solution
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland