Merck
  • Acute hemolytic anemia as an initial presentation of Wilson disease in children.
Acute hemolytic anemia as an initial presentation of Wilson disease in children.
Journal of pediatric hematology/oncology (2014-03-01)
Mona S El Raziky, Amal Ali, Amira El Shahawy, Mona M Hamdy
ABSTRACT
Wilson disease (WD) is an inherited disorder of copper metabolism. Hemolytic anemia in WD occurs in up to 17% of patients at some point during their illness. To screen for WD among children presenting with hemolytic anemia. Twenty cases (mean age, 8.8 ± 3.9 y) with Coombs-negative hemolytic anemia, attending the hematology clinic of children hospital, Cairo University, were screened for WD by serum ceruloplasmin level, 24 hours urinary copper before and after D-penicillamine challenge test, and slit-lamp examination for detecting Kayser-Fleischer rings. No case had low ceruloplasmin, whereas bilateral Kayser-Fleischer rings was detected in 5% of our cases. Urinary copper was elevated in 5% before and in 40% after D-penicillamine challenge test. According to the scoring system used, 1 case had definite WD and 7 cases were likely to have WD. These 8 (40%) cases were referred to as group B. Group B had a significantly lower hemoglobin, mean corpuscular volume, mean corpuscular hemoglobin, and reticulocytes (P=0.04, 0.001, 0.04, and 0.04, respectively) and a significantly higher urinary copper after penicillamine (P=0.000) when compared with group A (unlikely WD). WD is not uncommon in children with hemolytic anemia after exclusion of other common causes.
Sigma-Aldrich
D-Penicillamine, 98-101%
Penicillamine, European Pharmacopoeia (EP) Reference Standard