Merck
  • Preparative separation and purification of rebaudioside a from steviol glycosides using mixed-mode macroporous adsorption resins.
Preparative separation and purification of rebaudioside a from steviol glycosides using mixed-mode macroporous adsorption resins.
Journal of agricultural and food chemistry (2011-08-02)
Yongfeng Liu, Duolong Di, Qingqing Bai, Jintian Li, Zhenbin Chen, Song Lou, Helin Ye
ABSTRACT
Preparative separation and purification of rebaudioside A from steviol glycosides using mixed-mode macroporous adsorption resins (MARs) were systematically investigated. Mixed-mode MARs were prepared by a physical blending method. By evaluation of the adsorption/desorption ratio and adsorption/desorption capacity of mixed-mode MARs with different proportions toward RA and ST, the mixed-mode MAR 18 was chosen as the optimum strategy. On the basis of the static tests, it was found that the experimental data fitted best to the pseudosecond-order kinetics and Temkin-Pyzhev isotherm. Furthermore, the dynamic adsorption/desorption experiments were performed on the mini column packed with mixed-mode MAR 18. After one run treatment, the purity of rebaudioside A in purified product increased from 40.77 to 60.53%, with a yield rate of 38.73% (W/W), and that in residual product decreased from 40.77 to 36.17%, with a recovery yield of 57.61% (W/W). The total recovery yield reached 96.34% (W/W). The results showed that this method could be utilized in large-scale production of rebaudioside A from steviol glycosides in industry.
Sigma-Aldrich
Rebaudioside A, ≥96% (HPLC)
Sigma-Aldrich
Steviol hydrate, ≥98% (HPLC), powder