Merck
  • The function of ER-phagy receptors is regulated through phosphorylation-dependent ubiquitination pathways.
The function of ER-phagy receptors is regulated through phosphorylation-dependent ubiquitination pathways.
Nature communications (2023-12-16)
Rayene Berkane, Hung Ho-Xuan, Marius Glogger, Pablo Sanz-Martinez, Lorène Brunello, Tristan Glaesner, Santosh Kumar Kuncha, Katharina Holzhüter, Sara Cano-Franco, Viviana Buonomo, Paloma Cabrerizo-Poveda, Ashwin Balakrishnan, Georg Tascher, Koraljka Husnjak, Thomas Juretschke, Mohit Misra, Alexis González, Volker Dötsch, Paolo Grumati, Mike Heilemann, Alexandra Stolz
ABSTRACT
Selective autophagy of the endoplasmic reticulum (ER), known as ER-phagy, is an important regulator of ER remodeling and essential to maintain cellular homeostasis during environmental changes. We recently showed that members of the FAM134 family play a critical role during stress-induced ER-phagy. However, the mechanisms on how they are activated remain largely unknown. In this study, we analyze phosphorylation of FAM134 as a trigger of FAM134-driven ER-phagy upon mTOR (mechanistic target of rapamycin) inhibition. An unbiased screen of kinase inhibitors reveals CK2 to be essential for FAM134B- and FAM134C-driven ER-phagy after mTOR inhibition. Furthermore, we provide evidence that ER-phagy receptors are regulated by ubiquitination events and that treatment with E1 inhibitor suppresses Torin1-induced ER-phagy flux. Using super-resolution microscopy, we show that CK2 activity is essential for the formation of high-density FAM134B and FAM134C clusters. In addition, dense clustering of FAM134B and FAM134C requires phosphorylation-dependent ubiquitination of FAM134B and FAM134C. Treatment with the CK2 inhibitor SGC-CK2-1 or mutation of FAM134B and FAM134C phosphosites prevents ubiquitination of FAM134 proteins, formation of high-density clusters, as well as Torin1-induced ER-phagy flux. Therefore, we propose that CK2-dependent phosphorylation of ER-phagy receptors precedes ubiquitin-dependent activation of ER-phagy flux.
Sigma-Aldrich
Anti-FAM134B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-FAM134C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution