Merck
  • Spinal Irisin Gene Delivery Attenuates Burn Injury-Induced Muscle Atrophy by Promoting Axonal Myelination and Innervation of Neuromuscular Junctions.
Spinal Irisin Gene Delivery Attenuates Burn Injury-Induced Muscle Atrophy by Promoting Axonal Myelination and Innervation of Neuromuscular Junctions.
International journal of molecular sciences (2022-12-24)
Sheng-Hua Wu, I-Cheng Lu, Shih-Ming Yang, Chia-Fang Hsieh, Chee-Yin Chai, Ming-Hong Tai, Shu-Hung Huang
ABSTRACT
Muscle loss and weakness after a burn injury are typically the consequences of neuronal dysregulation and metabolic change. Hypermetabolism has been noted to cause muscle atrophy. However, the mechanism underlying the development of burn-induced motor neuropathy and its contribution to muscle atrophy warrant elucidation. Current therapeutic interventions for burn-induced motor neuropathy demonstrate moderate efficacy and have side effects, which limit their usage. We previously used a third-degree burn injury rodent model and found that irisin-an exercise-induced myokine-exerts a protective effect against burn injury-induced sensory and motor neuropathy by attenuating neuronal damage in the spinal cord. In the current study, spinal irisin gene delivery was noted to attenuate burn injury-induced sciatic nerve demyelination and reduction of neuromuscular junction innervation. Spinal overexpression of irisin leads to myelination rehabilitation and muscular innervation through the modulation of brain-derived neurotrophic factor and glial-cell-line-derived neurotrophic factor expression along the sciatic nerve to the muscle tissues and thereby modulates the Akt/mTOR pathway and metabolic derangement and prevents muscle atrophy.
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-p75NTR Antibody, ICD, serum, from rabbit
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture