Merck
  • Generation of Marmoset Monkey iPSCs with Self-Replicating VEE-mRNAs in Feeder-Free Conditions.
Generation of Marmoset Monkey iPSCs with Self-Replicating VEE-mRNAs in Feeder-Free Conditions.
Methods in molecular biology (Clifton, N.J.) (2021-03-19)
Stoyan G Petkov, Rüdiger Behr
ABSTRACT
The generation and culture of transgene-free induced pluripotent stem cells (iPSCs) from the common marmoset (Callithrix jacchus) present unique challenges due to the fact that the protocols developed for culture of human or mouse pluripotent cells are not sufficiently optimized for this particular monkey species. Here, we describe the procedures for the reprogramming of marmoset fetal fibroblasts to pluripotency with self-replicating mRNAs using a two-step approach, where intermediate primary colonies generated in the first reprogramming step are converted in the second step to iPSCs with customized marmoset culture medium. The resulting iPSCs are free of transgenes and can be maintained in long-term culture in feeder-free conditions.
Sigma-Aldrich
GSK-3 Inhibitor XVI, GSK-3 Inhibitor XVI - CAS 252917-06-9, is a cell-permeable, potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC₅₀ = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively).
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture