Merck
  • A Novel Kinase Inhibitor AX-0085 Inhibits Interferon-γ-Mediated Induction of PD-L1 Expression and Promotes Immune Reaction to Lung Adenocarcinoma Cells.
A Novel Kinase Inhibitor AX-0085 Inhibits Interferon-γ-Mediated Induction of PD-L1 Expression and Promotes Immune Reaction to Lung Adenocarcinoma Cells.
Cells (2022-01-12)
Jusong Kim, Haeyeon Jang, Gyu Jin Lee, Yelim Hur, Juhee Keum, Jung Ki Jo, Si-Eun Yun, Sung Jun Park, Young Jun Park, Myeong Jun Choi, Kye-Seong Kim, Jaesang Kim
ABSTRACT
In this study, we describe a novel kinase inhibitor AX-0085 which can suppress the induction of PD-L1 expression by Interferon-γ (IFN-γ) in lung adenocarcinoma (LUAD) cells. AX-0085 effectively blocks JAK2/STAT1 signaling initiated by IFN-γ treatment and prevents nuclear localization of STAT1. Importantly, we demonstrate that AX-0085 reverses the IFN-γ-mediated repression of T cell activation in vitro and enhances the anti-tumor activity of anti-PD-1 antibody in vivo when used in combination. Finally, transcriptomic analyses indicated that AX-0085 is highly specific in targeting the IFN-γ-pathway, thereby raising the possibility of applying this reagent in combination therapy with checkpoint inhibitor antibodies. It may be particularly relevant in cases in which PD-L1-mediated T cell exhaustion leads to immunoevasive phenotypes.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-ALPHA-TUBULIN antibody produced in rabbit, affinity isolated antibody