Merck
  • The invasive proteome of glioblastoma revealed by laser-capture microdissection.
The invasive proteome of glioblastoma revealed by laser-capture microdissection.
Neuro-oncology advances (2020-07-10)
Thomas Daubon, Joris Guyon, Anne-Aurélie Raymond, Benjamin Dartigues, Justine Rudewicz, Zakaria Ezzoukhry, Jean-William Dupuy, John M J Herbert, Frédéric Saltel, Rolf Bjerkvig, Macha Nikolski, Andreas Bikfalvi
ABSTRACT
Glioblastomas are heterogeneous tumors composed of a necrotic and tumor core and an invasive periphery. Here, we performed a proteomics analysis of laser-capture micro-dissected glioblastoma core and invasive areas of patient-derived xenografts. Bioinformatics analysis identified enriched proteins in central and invasive tumor areas. Novel markers of invasion were identified, the genes proteolipid protein 1 (PLP1) and Dynamin-1 (DNM1), which were subsequently validated in tumors and by functional assays. In summary, our results identify new networks and molecules that may play an important role in glioblastoma development and may constitute potential novel therapeutic targets.
Sigma-Aldrich
Anti-PLP1 (ab1) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.