Merck
  • Symmetric Mixed Sulfur-Selenium Fused Ring Systems as Potential Materials for Organic Field-Effect Transistors.
Symmetric Mixed Sulfur-Selenium Fused Ring Systems as Potential Materials for Organic Field-Effect Transistors.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-11-16)
Brigitte Holzer, Barbara Dellago, Ann-Katrin Thamm, Thomas Mathis, Berthold Stöger, Ernst Horkel, Christian Hametner, Bertram Batlogg, Johannes Fröhlich, Daniel Lumpi
ABSTRACT
A reliable synthetic protocol toward a series of fused chalcogenopheno[1]benzochalcogenophene (CBC) building blocks was developed based on a Fiesselmann reaction. The obtained CBC units were applied in McMurry and Stille coupling reactions toward symmetric regioisomeric ene-linked dimers. These π-conjugated compounds were characterized regarding their photophysical and electrochemical properties and proved to be materials with reduced HOMO-LUMO gaps compared to their sulfur-based analogues. Single-crystal X-ray diffraction experiments revealed strong intermolecular selenium-selenium and selenium-carbon interactions depending on the position and number of incorporated selenium atoms. Good field-effect transistor performance with charge carrier mobilities up to 4×10-3  cm2  V-1  s-1 and high on/off ratios could be observed.
Sigma-Aldrich
2′-Chloroacetophenone, 97%
Sigma-Aldrich
trans-1,2-Bis(tributylstannyl)ethene, 97%