Skip to Content
Merck
4-Deoxynivalenol in acetonitrile IRMM®, certified reference material

Deoxynivalenol solution

4-Deoxynivalenol in acetonitrile IRMM®, certified reference material
Synonym(s):
Deoxynivalenol solution, 3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, DON, Vomitoxin
Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique