
Calcium oxalate monohydrate

Synonym(s):
Oxalic acid calcium salt
Linear Formula:
(COO)2Ca · H2O
CAS Number:
Molecular Weight:
146.11
Products
Formula Weight
Manufacturer
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing