
Anti-HA−FITC antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-HA, Anti-HA, Anti-Influenza Hemagglutinin
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing