Merck
  • High constitutive glucocorticoid receptor beta in human neutrophils enables them to reduce their spontaneous rate of cell death in response to corticosteroids.
High constitutive glucocorticoid receptor beta in human neutrophils enables them to reduce their spontaneous rate of cell death in response to corticosteroids.
The Journal of experimental medicine (2001-03-10)
I Strickland, K Kisich, P J Hauk, A Vottero, G P Chrousos, D J Klemm, D Y Leung
ABSTRACT
Neutrophils are markedly less sensitive to glucocorticoids than T cells, making it difficult to control inflammation in neutrophil-mediated diseases. Development of new antiinflammatory strategies for such diseases would be aided by an understanding of mechanisms underlying differential steroid responsiveness. Two protein isoforms of the human glucocorticoid receptor (GR) exist, GRalpha and GRbeta, which arise from alternative splicing of the GR pre-mRNA primary transcripts. GRbeta does not bind glucocorticoids and is an inhibitor of GRalpha activity. Relative amounts of these two GRs can therefore determine the level of glucocorticoid sensitivity. In this study, human neutrophils and peripheral blood mononuclear cells (PBMCs) were studied to determine the relative amounts of each GR isoform. The mean fluorescence intensity (MFI) using immunofluorescence analysis for GRalpha was 475 +/- 62 and 985 +/- 107 for PBMCs and neutrophils, respectively. For GRbeta, the MFI was 350 +/- 60 and 1,389 +/- 143 for PBMCs and neutrophils, respectively (P < 0.05). After interleukin (IL)-8 stimulation of neutrophils, there was a statistically significant increase in intensity of GRbeta staining to 2,497 +/- 140 (P < 0.05). No change in GRalpha expression was observed. This inversion of the GRalpha/GRbeta ratio in human neutrophils compared with PBMCs was confirmed by quantitative Western analysis. Increased GRbeta mRNA expression in neutrophils at baseline, and after IL-8 exposure, was observed using RNA dot blot analysis. Increased levels of GRalpha/GRbeta heterodimers were found in neutrophils as compared with PBMCs using coimmunoprecipitation/Western analysis. Transfection of mouse neutrophils, which do not contain GRbeta, resulted in a significant reduction in the rate of cell death when treated with dexamethasone.We conclude that high constitutive expression of GRbeta by human neutrophils may provide a mechanism by which these cells escape glucocorticoid-induced cell death. Moreover, upregulation of this GR by proinflammatory cytokines such as IL-8 further enhances their survival in the presence of glucocorticoids during inflammation.
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)