Merck
  • [The use of iodobromine water on patients with periodontitis at a health resort].
[The use of iodobromine water on patients with periodontitis at a health resort].
Stomatologiia (1990-05-01)
L V Egorov
ABSTRACT
The technique of iodobromine water irrigations of the oral cavity in periodontitis has been developed to be used at health resorts of Anapa. A total of 4650 patients aged 25 to 52 with periodontitis of varying severity were treated; mineral water irrigations were preceded by removal of dental deposit. The treatment efficacy was assessed by functional methods: electrothermometry and rheography.
Sigma-Aldrich
Iodine monobromide, 98%