Merck
Diastereodivergent addition of allenylzincs to aryl glyoxylates.
Organic letters (2009-05-09)
Thomas Hameury, Jérôme Guillemont, Luc Van Hijfte, Véronique Bellosta, Janine Cossy
ABSTRACT
A diastereodivergent addition of allenylzincs to aryl glyoxylates was observed depending on the method used for the preparation of the allenylzinc reagent. The allenylzincs were prepared from propargylic benzoates in the presence of a palladium catalyst or by metalation of alkynes.
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, puriss., anhydrous, ≥98%