Merck
  • Cockayne syndrome group A and B proteins function in rRNA transcription through nucleolin regulation.
Cockayne syndrome group A and B proteins function in rRNA transcription through nucleolin regulation.
Nucleic acids research (2020-01-24)
Mustafa N Okur, Jong-Hyuk Lee, Wasif Osmani, Risako Kimura, Tyler G Demarest, Deborah L Croteau, Vilhelm A Bohr
ABSTRACT
Cockayne Syndrome (CS) is a rare neurodegenerative disease characterized by short stature, accelerated aging and short lifespan. Mutations in two human genes, ERCC8/CSA and ERCC6/CSB, are causative for CS and their protein products, CSA and CSB, while structurally unrelated, play roles in DNA repair and other aspects of DNA metabolism in human cells. Many clinical and molecular features of CS remain poorly understood, and it was observed that CSA and CSB regulate transcription of ribosomal DNA (rDNA) genes and ribosome biogenesis. Here, we investigate the dysregulation of rRNA synthesis in CS. We report that Nucleolin (Ncl), a nucleolar protein that regulates rRNA synthesis and ribosome biogenesis, interacts with CSA and CSB. In addition, CSA induces ubiquitination of Ncl, enhances binding of CSB to Ncl, and CSA and CSB both stimulate the binding of Ncl to rDNA and subsequent rRNA synthesis. CSB and CSA also increase RNA Polymerase I loading to the coding region of the rDNA and this is Ncl dependent. These findings suggest that CSA and CSB are positive regulators of rRNA synthesis via Ncl regulation. Most CS patients carry mutations in CSA and CSB and present with similar clinical features, thus our findings provide novel insights into disease mechanism.
Product Description
Millipore
Benzonase® Nuclease, Purity > 99%, Effective viscosity reduction and removal of nucleic acids from protein solutions
Sigma-Aldrich
Anti-Nucleolin Antibody, clone 3G4B2, culture supernatant, clone 3G4B2, Upstate®