Merck
  • MeCP2-regulated miRNAs control early human neurogenesis through differential effects on ERK and AKT signaling.
MeCP2-regulated miRNAs control early human neurogenesis through differential effects on ERK and AKT signaling.
Molecular psychiatry (2017-04-26)
N Mellios, D A Feldman, S D Sheridan, J P K Ip, S Kwok, S K Amoah, B Rosen, B A Rodriguez, B Crawford, R Swaminathan, S Chou, Y Li, M Ziats, C Ernst, R Jaenisch, S J Haggarty, M Sur
ABSTRACT
Rett syndrome (RTT) is an X-linked, neurodevelopmental disorder caused primarily by mutations in the methyl-CpG-binding protein 2 (MECP2) gene, which encodes a multifunctional epigenetic regulator with known links to a wide spectrum of neuropsychiatric disorders. Although postnatal functions of MeCP2 have been thoroughly investigated, its role in prenatal brain development remains poorly understood. Given the well-established importance of microRNAs (miRNAs) in neurogenesis, we employed isogenic human RTT patient-derived induced pluripotent stem cell (iPSC) and MeCP2 short hairpin RNA knockdown approaches to identify novel MeCP2-regulated miRNAs enriched during early human neuronal development. Focusing on the most dysregulated miRNAs, we found miR-199 and miR-214 to be increased during early brain development and to differentially regulate extracellular signal-regulated kinase (ERK)/mitogen-activated protein kinase and protein kinase B (PKB/AKT) signaling. In parallel, we characterized the effects on human neurogenesis and neuronal differentiation brought about by MeCP2 deficiency using both monolayer and three-dimensional (cerebral organoid) patient-derived and MeCP2-deficient neuronal culture models. Inhibiting miR-199 or miR-214 expression in iPSC-derived neural progenitors deficient in MeCP2 restored AKT and ERK activation, respectively, and ameliorated the observed alterations in neuronal differentiation. Moreover, overexpression of miR-199 or miR-214 in the wild-type mouse embryonic brains was sufficient to disturb neurogenesis and neuronal migration in a similar manner to Mecp2 knockdown. Taken together, our data support a novel miRNA-mediated pathway downstream of MeCP2 that influences neurogenesis via interactions with central molecular hubs linked to autism spectrum disorders.
Sigma-Aldrich
Anti-Sox1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-PAX6 Antibody, from rabbit, purified by affinity chromatography