
Thiamine Thiocarbamate

Synonym(s):
(3RS)-3-[[[(4-Amino-2-methylpyrimidin-5-yl)methyl]thiocarbamoyl]sulfanyl]-4-oxopentyl acetate (ketodithiocarbamate)
Empirical Formula (Hill Notation):
C14H20N4O3S2
CAS Number:
Molecular Weight:
356.46
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing