
Icariside II

Synonym(s):
Baohuoside I, Icariin II
Empirical Formula (Hill Notation):
C27H30O10
CAS Number:
Molecular Weight:
514.52
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing