
2-Deoxy-D-glucose 6-phosphate sodium salt

Synonym(s):
2-Deoxy-D-glucose 6-phosphate sodium salt
Empirical Formula (Hill Notation):
C6H13O8P · xNa+
CAS Number:
Molecular Weight:
244.14 (free acid basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing