
2-Amino-3-(1H-1,2,4-triazol-1-yl)propanoic acid

Synonym(s):
α-Amino-1H-1,2,4-triazole-1-propanoic acid, 3-(1,2,4-Triazol-1-yl)-DL-alanine, H-β-(1,2,4-Triazol-1-yl)-DL-Ala-OH
Empirical Formula (Hill Notation):
C5H8N4O2
CAS Number:
Molecular Weight:
156.14
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1