HS120598
Mini Vortex Mixer
AC/DC input 100 V / 240 V, universal plug set, gray-blue
All Photos(2)
About This Item
UNSPSC Code:
41103814
NACRES:
NB.13
Recommended Products
material
gray-blue
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120598
General description
Simple, touch start operation offers exceptional mixing performance with one hand
- Low profile design and small footprint (only 4.3” wide) saves valuable space
- Made of ABS and silicone materials that resist most commonly used chemicals
- Stable platform and elastomer feet ensure unit stays stable during operation
- When performing repetitive procedures, a low activation force helps prevent user fatigue
Packaging
INCLUDES:
1 Mini Vortexer
2 Silicone Caps
1 Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs
1 Mini Vortexer
2 Silicone Caps
1 Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs