HPLC Analysis of Peptides and Proteins on Proteomix® SCX-NP1.7, 3 cm x 4.6 mm I.D. Column
Materials
analytical column
Product No.
Description
Pricing
Proteomix® SCX-NP1.7 Strong Cation Exchange Column
L × I.D. 3 cm × 4.6 mm, 1.7 μm particle size, stainless steelstandard
Product No.
Description
Pricing
Ribonuclease A from bovine pancreas
Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg proteinCONDITIONS
column
Proteomix SCX-NP1.7, 3 cm x 4.6 mm I.D., 1.7 μm particles (Z777145)
mobile phase
(A) 20 mM sodium phosphate, pH 6.0; (B) A + 1.0 M NaCl
gradient
0 to 50% B in 5 min; 15 min prewash
flow rate
2 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
5 μL
sample
1 mg/mL
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies