Merck
  • Elevated expression of TANK-binding kinase 1 enhances tamoxifen resistance in breast cancer.
Elevated expression of TANK-binding kinase 1 enhances tamoxifen resistance in breast cancer.
Proceedings of the National Academy of Sciences of the United States of America (2014-01-23)
Congwen Wei, Yuan Cao, Xiaoli Yang, Zirui Zheng, Kai Guan, Qiang Wang, Yanhong Tai, Yanhong Zhang, Shengli Ma, Ye Cao, Xiaoxing Ge, Changzhi Xu, Jia Li, Hui Yan, Youguo Ling, Ting Song, Lin Zhu, Buchang Zhang, Quanbin Xu, Chengjin Hu, Xiu-wu Bian, Xiang He, Hui Zhong
ABSTRACT
Resistance to antiestrogens is one of the major challenges in breast cancer treatment. Although phosphorylation of estrogen receptor α (ERα) is an important factor in endocrine resistance, the contributions of specific kinases in endocrine resistance are still not fully understood. Here, we report that an important innate immune response kinase, the IκB kinase-related TANK-binding kinase 1 (TBK1), is a crucial determinant of resistance to tamoxifen therapies. We show that TBK1 increases ERα transcriptional activity through phosphorylation modification of ERα at the Ser-305 site. Ectopic TBK1 expression impairs the responsiveness of breast cancer cells to tamoxifen. By studying the specimens from patients with breast cancer, we find a strong positive correlation of TBK1 with ERα, ERα Ser-305, and cyclin D1. Notably, patients with tumors highly expressing TBK1 respond poorly to tamoxifen treatment and show high potential for relapse. Therefore, our findings suggest that TBK1 contributes to tamoxifen resistance in breast cancer via phosphorylation modification of ERα.
Sigma-Aldrich
Anti-c-Myc−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-IKKε Antibody, Upstate®, from rabbit