Merck
Complex genetics of drug resistance in Mycobacterium tuberculosis.
Nature genetics (2013-09-28)
Digby F Warner, Valerie Mizrahi
ABSTRACT
Three new studies have used whole-genome sequencing of M. tuberculosis to demonstrate unexpected complexity in the modern evolution of drug-resistant tuberculosis, and a fourth study suggests a close evolutionary relationship between the pathogen and its human host over a period of 70,000 years. Collectively, the observations in these studies suggest that future strategies to tackle drug-resistant tuberculosis must integrate host genetics with detailed strain epidemiology.
Sigma-Aldrich
Ethambutol dihydrochloride, antimycobacterial