Merck
  • Spectral studies and structure of a 2-hydroxyacetophenone 3-hexamethyleneiminyl thiosemicarbazonate(-2) copper(II) complex containing 1,10-phenanthroline.
Spectral studies and structure of a 2-hydroxyacetophenone 3-hexamethyleneiminyl thiosemicarbazonate(-2) copper(II) complex containing 1,10-phenanthroline.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-03-28)
Rohith P John, A Sreekanth, Maliyeckal R Prathapachandra Kurup, Anwar Usman, Abdul Razak Ibrahim, Hoong-Kun Fun
ABSTRACT
The spectral studies and structure of a ternary complex of copper(II) with 2-hydroxyacetophenone 3-hexamethyliminylthiosemicarbazonate (L(2-)) and 1,10-phenanthroline (phen) are reported. The thiosemicarbazone binds to the metal as a dianionic ONS-donor (L(2-)) ligand, and forms a complex of the stoichiometry [CuLphen]. The copper(II) complex was characterized by IR and UV/Vis spectroscopies, as well as by solid state room-temperature magnetic susceptibility. Spin Hamiltonian and bonding parameters of the compound are calculated from the EPR spectra. Computer simulation of EPR spectrum in DMF at 77 K aided the calculation of magnetic and bonding parameters of the compound. The structure of the compound is solved by single crystal X-ray diffraction. The geometry around copper is distorted square pyramidal.
Sigma-Aldrich
2′-Hydroxyacetophenone, ≥98%
Sigma-Aldrich
2′-Hydroxyacetophenone, ReagentPlus®, 99%