Merck
  • Hydrogen Bonding-Induced Assembled Structures and Photoresponsive Behavior of Azobenzene Molecule/Polyethylene Glycol Complexes.
Hydrogen Bonding-Induced Assembled Structures and Photoresponsive Behavior of Azobenzene Molecule/Polyethylene Glycol Complexes.
Polymers (2019-08-21)
Hsin-Tzu Tai, Yen-Chun Lin, Jing-Yao Ma, Chieh-Tsung Lo
ABSTRACT
We investigated the self-assembled structures and photoresponsive and crystallization behaviors of supramolecules composed of 4-methoxy-4'-hydroxyazobenzene (Azo) molecules and polyethylene glycol (PEG) that were formed through hydrogen-bonding interactions. The Azo/PEG complexes exhibited the characteristics of photoresponse and crystallization, which originated from Azo and PEG, respectively. When Azo/PEG complexes were dissolved in solvents, hydrogen-bonding interaction hindered the rotation and inversion of mesogens, causing a reduction in the photoisomerization rate compared with the photoisomerization rate of the neat Azo. The confinement of Azo/PEG complexes in thin films further resulted in a substantial decrease in the photoisomerization rate but an increase in the amounts of H-aggregated and J-aggregated mesogens. Regarding PEG crystallization, ultraviolet irradiation of Azo/PEG complexes increased the quantity of high-polarity cis isomers, which improved the compatibility between mesogens and PEG, subsequently increasing the crystallization temperature of PEG. Moreover, the complexation of Azo and PEG induced microphase separation, forming a lamellar morphology. Within the Azo-rich microphases, mesogens aggregated to form tilted monosmectic layers. By contrast, PEG crystallization within the PEG-rich microphases was hard confined, indicating that the domain size of the lamellar morphology was unchanged during PEG crystallization.
Sigma-Aldrich
p-Anisidine, 99%
Sigma-Aldrich
Sodium nitrite, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%