Merck
  • Etidronate attenuates tactile allodynia by spinal ATP release inhibition in mice with partial sciatic nerve ligation.
Etidronate attenuates tactile allodynia by spinal ATP release inhibition in mice with partial sciatic nerve ligation.
Naunyn-Schmiedeberg's archives of pharmacology (2018-12-06)
Ryota Yamagata, Wataru Nemoto, Osamu Nakagawasai, Wan-Yi Hung, Kazuhiro Shima, Yasuo Endo, Koichi Tan-No
ABSTRACT
Etidronate is widely used as a therapeutic agent for osteoporosis. We have recently shown that intrathecal administration of etidronate into mice produces an analgesic effect against the capsaicin-induced nociceptive behavior. However, the effect of etidronate on neuropathic pain at the spinal level remains unknown. Therefore, we examined whether etidronate attenuates pain after partial sciatic nerve ligation (PSNL). We evaluated tactile allodynia 7 days after PSNL by measuring paw withdrawal with the von Frey filament test. The mRNA and protein levels of SLC17A9 in the ipsilateral lumbar dorsal spinal cord of PSNL-operated mice were determined using real-time PCR and western blotting, respectively. PSNL-induced tactile allodynia was attenuated by oral and intrathecal administration of etidronate, with maximum efficiency at 90 and 60 min after injection, respectively. The anti-allodynic effect of intrathecally administered etidronate was completely inhibited by an intrathecal administration of adenosine triphosphate (ATP). The solute carrier family, SLC17, mediates the transport of pain transmitters, like ATP and glutamate. Indeed, we detected several members of the SLC17 family in the mouse dorsal lumbar spinal cord. Among the detected mRNAs, only Slc17a9, encoding for neuronal vesicular ATP transporter, was significantly increased upon PSNL. SLC17A9 protein levels were also significantly increased. In mice subjected to PSNL, SLC17A9 was present in neurons and microglia, but not in astrocytes of the lumbar superficial dorsal horn. Collectively, our results suggest that etidronate produces its anti-allodynic effects by inhibiting SLC17A9-dependent exocytotic ATP release from the dorsal horn in mice subjected to PSNL.