
Salmeterol Related Compound H

Synonym(s):
1-Hydroxy-4-[2-hydroxy-5-(1-hydroxy-2-{[6-(4-phenylbutoxy)hexyl]amino}ethyl)benzyl]-2-naphthoic acid
Empirical Formula (Hill Notation):
C36H43NO6
CAS Number:
Molecular Weight:
585.73
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing