Skip to Content
Merck
vibrant-m

Acebutolol Related Compound I

vibrant-m
Synonym(s):
N-[3-Acetyl-4-[(2RS)-3-(ethylamino)-2-hydroxypropoxy]phenyl]butanamide
Empirical Formula (Hill Notation):
C17H26N2O4
CAS Number:
Molecular Weight:
322.40

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1