
4-Vinyl-1,3-dioxolan-2-one

Synonym(s):
VEC, Vinyl ethylene carbonate
Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
Products
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing