Merck
HPLC Analysis of PAHs in Canola Oil on SUPELCOSIL LC-PAH after SPE using Supelclean EZ-POP NP
HPLC Analysis of PAHs in Canola Oil on SUPELCOSIL™ LC-PAH after SPE using Supelclean™ EZ-POP NP application for SPE, application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-PAH HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
SPE tube or plate
Product No.
Description
Pricing
Supelclean EZ-POP NP
volume 12 mL, pkg of 20
standard
Product No.
Description
Pricing
PAH Calibration Mix
TraceCERT®, certified reference material, 10 μg/mL each component in acetonitrile
Polynuclear Aromatic Hydrocarbons Mix
certified reference material, 2000 μg/mL each component in benzene: dichloromethane (50:50), ampule of 1 mL
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
Canola oil spiked with PAHs at 10 ng/g
SPE tube/cartridge
Supelclean EZ-POP NP, 12 mL, pk of 20 (54341-U)
condition
10 mL acetone, dry under 10-15" mercury for 10 minutes
sample addition
0.5 mL weighed accurately onto cartridge
elution
15 mL acetonitrile
eluate post-treatment
concentrate to final volume of 0.5mL at 40 °C under nitrogen
column
SUPELCOSIL LC-PAH 25 cm x 4.6 mm I.D., 5 μm particles (58229)
mobile phase
[A] water [B] acetonitrile
gradient
40% B held for 5 min; to 100% B in 15 min; held at 100% B for 12 min
flow rate
2.4 mL/min
pressure
2790 psi at start (192 bar)
column temp.
25 °C
detector
Fluorescence, programmed.
Description
Analysis Note
PAHs are ubiquitous in the environment, resulting from both natural and manmade sources of combustion. They can contaminate food crops through environmental exposure to particulates, automobile exhaust, etc. In the case of edible oils, additional PAHs can be introduced during actual production processes such as seed drying [1]. In this application, EZ-POP NP SPE is used to extract PAHs from soybean oil. The 16 PAHs designated in many US EPA methods (less acenaphthylene) were analyzed by HPLC-FLD after extraction from a spiked sample of oil.
Other Notes
Detector: Naphthalene, Acenaphthene, Fluorene - 225nm/320nm; Phenanthrene, Anthracene - 250nm/368nm; Fluoranthene, Pyrene - 237nm/440nm; Benzo[a]anthracene, Chrysene - 265nm/380nm; Benzo[b]fluoranthene, Benzo[k]fluoranthene, Benzo[a]pyrene, Dibenzo[a,h]anthracene - 280nm/420nm; Benzo[g,h,i]perylene, Indeno[1,2,3-cd]pyrene, Benzo[b]chrysene - 300nm/466nm
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC