Merck
  • Repeated AAV-mediated gene transfer by serotype switching enables long-lasting therapeutic levels of hUgt1a1 enzyme in a mouse model of Crigler-Najjar Syndrome Type I.
Repeated AAV-mediated gene transfer by serotype switching enables long-lasting therapeutic levels of hUgt1a1 enzyme in a mouse model of Crigler-Najjar Syndrome Type I.
Gene therapy (2017-08-15)
L Bočkor, G Bortolussi, A Iaconcig, G Chiaruttini, C Tiribelli, M Giacca, F Benvenuti, L Zentilin, A F Muro
ABSTRACT
Adeno-associated virus (AAV) -mediated gene therapy is a promising strategy to treat liver-based monogenic diseases. However, two major obstacles limit its success: first, vector dilution in actively dividing cells, such as hepatocytes in neonates/children, due to the non-integrating nature of the vector; second, development of an immune response against the transgene and/or viral vector. Crigler-Najjar Syndrome Type I is a rare monogenic disease with neonatal onset, caused by mutations in the liver-specific UGT1 gene, with toxic accumulation of unconjugated bilirubin in plasma, tissues and brain. To establish an effective and long lasting cure, we applied AAV-mediated liver gene therapy to a relevant mouse model of the disease. Repeated gene transfer to adults by AAV-serotype switching, upon neonatal administration, resulted in lifelong correction of total bilirubin (TB) levels in both genders. In contrast, vector loss over time was observed after a single neonatal administration. Adult administration resulted in lifelong TB levels correction in male, but not female Ugt1