Merck
  • Favipiravir inhibits acetaminophen sulfate formation but minimally affects systemic pharmacokinetics of acetaminophen.
Favipiravir inhibits acetaminophen sulfate formation but minimally affects systemic pharmacokinetics of acetaminophen.
British journal of clinical pharmacology (2015-03-27)
Yanli Zhao, Jerold S Harmatz, Carol R Epstein, Yukako Nakagawa, Chie Kurosaki, Tetsuro Nakamura, Takumi Kadota, Dennis Giesing, Michael H Court, David J Greenblatt
ABSTRACT
The antiviral agent favipiravir is likely to be co-prescribed with acetaminophen (paracetamol). The present study evaluated the possiblility of a pharmacokinetic interaction between favipiravir and acetaminophen, in vitro and in vivo. The effect of favipivir on the transformation of acetaminophen to its glucuronide and sulfate metabolites was studied using a pooled human hepatic S9 fraction in vitro. The effect of acute and extended adminstration of favipiravir on the pharmacokinetics of acetaminophen and metabolites was evaluated in human volunteers. Favipiravir inhibited the in vitro formation of acetaminophen sulfate, but not acetaminophen glucuronide. In human volunteers, both acute (1 day) and extended (6 days) administration of favipiravir slightly but significantly increased (by about 20 %) systemic exposure to acetaminophen (total AUC), whereas Cmax was not significantly changed. AUC for acetaminophen glucuronide was increased by 23 to 35 % above control by favipiravir, while AUC for acetaminophen sulfate was reduced by about 20 % compared to control. Urinary excretion of acetaminophen sulfate was likewise reduced to 44 to 65 % of control values during favipiravir co-administration, while excretion of acetaminophen glucuronide increased to 17 to 32 % above control. Favipiravir inhibits acetaminophen sulfate formation in vitro and in vivo. However the increase in systemic exposure to acetaminophen due to favipiravir co-administration, though statistically significant, is small in magnitude and unlikely to be of clinical importance.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
p-Acetamidophenyl β-D-glucuronide sodium salt, >98% (TLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Paracetamol sulfate potassium salt, solid, ≥97% (HPLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
3-Acetamidophenol, 97%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)