Merck
  • Efficient end-capping synthesis of neutral donor-acceptor [2]rotaxanes under additive-free and mild conditions.
Efficient end-capping synthesis of neutral donor-acceptor [2]rotaxanes under additive-free and mild conditions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-10-07)
Yuya Domoto, Shohei Sase, Kei Goto
ABSTRACT
Efficient end-capping synthesis of neutral donor-acceptor (D-A) [2]rotaxanes without loading any catalysts or activating agents was achieved by utilizing high reactivity of a pentacoordinated hydrosilane toward salicylic acid derivatives. As components of [2]rotaxanes, an electron-deficient naphthalenediimide-containing axle with a salicylic acid terminus and several electron-rich bis(naphthocrown) ether macrocycles were employed. End-capping reactions with the pentacoordinated hydrosilane underwent smoothly even at low temperature to afford the corresponding [2]rotaxanes in good yields. A [2]rotaxane containing bis-1,5-(dinaphtho)-38-crown-10 ether as a wheel molecule was synthesized and isolated in 84% yield by the end-capping at -10 °C, presenting the highest yield ever reported for the end-capping synthesis of a neutral D-A [2]rotaxane. It was found that the yields of the [2]rotaxanes in the end-capping reactions were almost parallel to the formation ratios of the corresponding pseudo[2]rotaxanes estimated by utilizing model systems. These results indicate that the end-capping reaction using the pentacoordinated hydrosilane proceeded without perturbing the threading process, and most of the pseudo[2]rotaxanes underwent efficient end-capping reaction even at low temperature.
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%
Supelco
3-Nitrobenzyl alcohol, suitable for mass spectrometry (MS), ≥99.5%