Merck
  • L-proline catalyzed asymmetric transfer aldol reaction between diacetone alcohol and aldehydes.
L-proline catalyzed asymmetric transfer aldol reaction between diacetone alcohol and aldehydes.
Chemical communications (Cambridge, England) (2004-10-30)
S Chandrasekhar, Ch Narsihmulu, N Ramakrishna Reddy, S Shameem Sultana
ABSTRACT
We demonstrate for the first time, L-proline as a chiral catalyst for transfer aldol reaction between aldehydes and diacetone alcohol.
Sigma-Aldrich
4-Hydroxy-4-methyl-2-pentanone, 99%
Pricing and availability is not currently available.