Merck
  • Extensive Chemometric Investigations of Distinctive Patterns and Levels of Biogenic Amines in Fermented Foods: Human Health Implications.
Extensive Chemometric Investigations of Distinctive Patterns and Levels of Biogenic Amines in Fermented Foods: Human Health Implications.
Foods (Basel, Switzerland) (2020-12-10)
Martin Grootveld, Benita C Percival, Jie Zhang
ABSTRACT
Although biogenic amines (BAs) present in fermented foods exert important health-promoting and physiological function support roles, their excessive ingestion can give rise to deleterious toxicological effects. Therefore, here we have screened the BA contents and supporting food quality indices of a series of fermented food products using a multianalyte-chemometrics strategy. A liquid chromatographic triple quadrupole mass spectrometric (LC-MS/MS) technique was utilized for the simultaneous multicomponent analysis of 8 different BAs, and titratable acidity, pH, total lipid content, and thiobarbituric acid-reactive substances (TBARS) values were also determined. Rigorous univariate and multivariate (MV) chemometric data analysis strategies were employed to evaluate results acquired. Almost all foods analyzed had individual and total BA contents that were within recommended limits. The chemometrics methods applied were useful for recognizing characteristic patterns of BA analytes and food quality measures between some fermented food classes, and for assessing their inter-relationships and potential metabolic sources. MV analysis of constant sum-normalized BA profile data demonstrated characteristic signatures for cheese (cadaverine only), fermented cod liver oil (2-phenylethylamine, tyramine, and tryptamine), and wine/vinegar products (putrescine, spermidine, and spermine). In conclusion, this LC-MS/MS-linked chemometrics approach was valuable for (1) contrasting and distinguishing BA catabolite signatures between differing fermented foods, and (2) exploring and evaluating the health benefits and/or possible adverse public health risks of such products.
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Tryptamine, ≥97%