Merck
  • Temporal and spatial transcriptomic and microRNA dynamics of CAM photosynthesis in pineapple.
Temporal and spatial transcriptomic and microRNA dynamics of CAM photosynthesis in pineapple.
The Plant journal : for cell and molecular biology (2017-07-04)
Ching M Wai, Robert VanBuren, Jisen Zhang, Lixian Huang, Wenjing Miao, Patrick P Edger, Won C Yim, Henry D Priest, Blake C Meyers, Todd Mockler, J Andrew C Smith, John C Cushman, Ray Ming
ABSTRACT
The altered carbon assimilation pathway of crassulacean acid metabolism (CAM) photosynthesis results in an up to 80% higher water-use efficiency than C3 photosynthesis in plants making it a potentially useful pathway for engineering crop plants with improved drought tolerance. Here we surveyed detailed temporal (diel time course) and spatial (across a leaf gradient) gene and microRNA (miRNA) expression patterns in the obligate CAM plant pineapple [Ananas comosus (L.) Merr.]. The high-resolution transcriptome atlas allowed us to distinguish between CAM-related and non-CAM gene copies. A differential gene co-expression network across green and white leaf diel datasets identified genes with circadian oscillation, CAM-related functions, and source-sink relations. Gene co-expression clusters containing CAM pathway genes are enriched with clock-associated cis-elements, suggesting circadian regulation of CAM. About 20% of pineapple microRNAs have diel expression patterns, with several that target key CAM-related genes. Expression and physiology data provide a model for CAM-specific carbohydrate flux and long-distance hexose transport. Together these resources provide a list of candidate genes for targeted engineering of CAM into C3 photosynthesis crop species.
Product Description
Supelco
Fructose Assay Kit, sufficient for 20 assays
Supelco
Sucrose Assay Kit, sufficient for 20 assays
Supelco
Glucose (HK) Assay Kit, sufficient for 20 assays
Supelco
Starch (HK) Assay Kit, sufficient for 20 assays