Merck
  • Lifetime methamphetamine dependence is associated with cerebral microgliosis in HIV-1-infected adults.
Lifetime methamphetamine dependence is associated with cerebral microgliosis in HIV-1-infected adults.
Journal of neurovirology (2016-04-22)
Virawudh Soontornniyomkij, Anya Umlauf, Benchawanna Soontornniyomkij, Isabella B Batki, David J Moore, Eliezer Masliah, Cristian L Achim
ABSTRACT
Methamphetamine (Meth) use is common among HIV-infected persons. It remains unclear whether Meth dependence is associated with long-lasting degenerative changes in the brain parenchyma and microvasculature of HIV-infected individuals. We examined the postmortem brains of 78 HIV-infected adults, twenty of whom were diagnosed with lifetime Meth dependence (18 past and two current at the final follow-up visit). Using logistic regression models, we analyzed associations of Meth with cerebral gliosis (immunohistochemistry for ionized calcium-binding adapter molecule-1 (Iba1) and glial fibrillary acidic protein (GFAP) in frontal, temporo-parietal, and putamen-internal capsule regions), synaptodendritic loss (confocal microscopy for synaptophysin (SYP) and microtubule-associated protein-2 (MAP2) in frontal cortex), β-amyloid plaque deposition (immunohistochemistry in frontal and temporo-parietal cortex and putamen), and arteriolosclerosis (histopathology in forebrain white matter). We found that Meth was associated with marked Iba1 gliosis in the temporo-parietal region (odds ratio, 4.42 (95 % confidence interval, 1.36, 14.39), p = 0.014, n = 62), which remained statistically significant after adjusting for HIV encephalitis, white matter lesions, and opportunistic diseases (n = 61); hepatitis C virus seropositivity (n = 54); and lifetime dependence on alcohol, opiates, and cannabis (n = 62). There was no significant association of Meth with GFAP gliosis, SYP or MAP2 loss, β-amyloid plaque deposition, or arteriolosclerosis. In conclusion, we found lifetime Meth dependence to be associated with focal cerebral microgliosis among HIV-infected adults, but not with other brain degenerative changes examined. Some of the changes in select brain regions might be reversible following extended Meth abstinence or, alternatively, might have not been induced by Meth initially.
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SY38, clone SY38, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAP2 Antibody, clone 5F9, clone 5F9, Upstate®, from mouse