Merck
  • Systemic network analysis identifies XIAP and IκBα as potential drug targets in TRAIL resistant BRAF mutated melanoma.
Systemic network analysis identifies XIAP and IκBα as potential drug targets in TRAIL resistant BRAF mutated melanoma.
NPJ systems biology and applications (2018-11-13)
Greta Del Mistro, Philippe Lucarelli, Ines Müller, Sébastien De Landtsheer, Anna Zinoveva, Meike Hutt, Martin Siegemund, Roland E Kontermann, Stefan Beissert, Thomas Sauter, Dagmar Kulms
ABSTRACT
Metastatic melanoma remains a life-threatening disease because most tumors develop resistance to targeted kinase inhibitors thereby regaining tumorigenic capacity. We show the 2nd generation hexavalent TRAIL receptor-targeted agonist IZI1551 to induce pronounced apoptotic cell death in mutBRAF melanoma cells. Aiming to identify molecular changes that may confer IZI1551 resistance we combined Dynamic Bayesian Network modelling with a sophisticated regularization strategy resulting in sparse and context-sensitive networks and show the performance of this strategy in the detection of cell line-specific deregulations of a signalling network. Comparing IZI1551-sensitive to IZI1551-resistant melanoma cells the model accurately and correctly predicted activation of NFκB in concert with upregulation of the anti-apoptotic protein XIAP as the key mediator of IZI1551 resistance. Thus, the incorporation of multiple regularization functions in logical network optimization may provide a promising avenue to assess the effects of drug combinations and to identify responders to selected combination therapies.
Sigma-Aldrich
Anti-NOXA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)