
Silybin (A + B mixture)

Synonym(s):
Silybin, (2R,3R)-3,5,7-Trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-hydroxymethyl-2,3-dihydrobenzo[1,4]dioxin-6-yl]chroman-4-one, Silibinin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing