
Cholecystokinin (CCK) Fragment 26-33 Amide, Non-sulfated

Synonym(s):
CCK-8, non-sulfated
Empirical Formula (Hill Notation):
C49H62N10O13S2
CAS Number:
Molecular Weight:
1063.21
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing