
Ceftazidime pentahydrate

Empirical Formula (Hill Notation):
C22H22N6O7S2 · 5H2O
CAS Number:
Molecular Weight:
636.65
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing